Learn More
Lenz Laborglasinstrumente™ Flat Bottom Hollow Stoppers, Hexagonal Plate
Description
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Description | Hollow |
| Quantity | 10/Pk. |
| Standard Taper Joint | Thread Size: 29/32 Cone NS |
| Material | Borosilicate Glass |
| Product Type | Hollow Stopper |