Learn More
Lenz Laborglasinstrumente™ Flat Bottom Hollow Stoppers, Hexagonal Plate
Click to view available options
Standard Taper Joint:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 34/35
Thread Size: 29/32 Cone NS
Unit Size:
Pack of 10
Pack of 5
Description
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Quantity | 10/Pk. |
| Standard Taper Joint | NS 10/19 |
| Material | Borosilicate Glass |
| Product Type | Hollow Stopper |