Learn More
Brand™ Haemocytometer Cover Glasses, Boro 3.3
Product Code. 10101731 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
24 mm
Width (Metric):
24 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
10101731 24 mm 24 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10101731 Supplier Brand™ Supplier No. 723014
Estimated Shipment: 11-12-2026
Manufactured from borosilicate glass and complies with DIN ISO 8255 standards. BRAND™ Borosilicate Glass Haemacytometer Cover Glasses differ from ordinary cover glasses by their plain ground and polished surface.
  • Borosilicate glass, DIN ISO 8255.
  • Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Flatness tolerance ± 3 μm.
  • They differ from ordinary cover glasses by their plain ground and polished surface.
  • Packing: 2 cover glasses in a plastic bag, separated by tissue paper.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hemacytometer Coverslip
Material Borosilicate Glass
Length (Metric) 24 mm
Width (Metric) 24 mm
Thickness (Metric) 0.4 mm
Dimensions (L x W) 24 x 24 mm