Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottles without Head
Description
- Acc. to DIN 12596, without head
- With socket NS 29/32
- Available in a range of capacities
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass |
| Type | Gas Washing Bottle without Head |
| Includes | Socket |
| Standard Taper Joint | NS 29/32 |
| Height (Metric) | 200 mm |
| Certifications/Compliance | DIN 12463 |
| Quantity | 1 Pack |