Learn More
BRAND™ Borosilicate Glass Funnel
Product Code. p-8003551 Shop All Brand Products
missing translation for 'orderingAttributeHoverText'
Diameter (Metric)Top:
35mm
40mm
50mm
55mm
60mm
70mm
80mm
100mm
120mm
150mm
Length (Metric) Stem:
35mm
40mm
50mm
55mm
60mm
70mm
80mm
100mm
120mm
150mm
missing translation for 'unitSize'
Each
Product Code. 10611672
missing translation for 'mfr': BRAND™ 145505
Only null left
Add to basket
Add to basket
Manufactured from borosilicate glass. BRAND™ Borosilicate Glass Funnel includes a ground or fire-polished upper rim. Short stem provides optimal filtration. Angled at 60°C.
  • Manufactured from borosilicate glass.
  • Ground or fire-polished upper rim.
  • Short stem provides optimal filtration.
  • Angled at 60°C.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 35mm
Diameter (Metric) Stem 6mm
Material Borosilicate Glass
Angle 60°
Length (Metric) Stem 35mm