Learn More
Quickfit™ Borosilicate Glass Digestion Tube with Socket
Product Code. 15597990
Change view
Click to view available options
Diameter (Metric) Outer:
16 mm
26 mm
42 mm
Overall Length (Metric):
145 mm
255 mm
300 mm
Unit Size:
Pack of 20
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Overall Length (Metric) Unit Size
15597990 26 mm 255 mm Pack of 20
12425426 16 mm 145 mm Pack of 5
12435426 42 mm 300 mm Pack of 5
15587990 26 mm 300 mm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15597990 Supplier Quickfit™ Supplier No. TJ75/26/3
  • ONLINE EXCLUSIVE PRICE
    7780.00 NOK
    6865.23 NOK / Pack of 20
    Save 914.77 NOK
    11% Off
Estimated Shipment: 03-03-2027
Includes
4 x Packs of 5
Designed for use with blockheaters.
  • Contains Quickfit precision-ground socket to accept air condensers
  • Manufactured from borosilicate glass for highest chemical and thermal durability
Specifications
Material Borosilicate Glass (3.3)
Diameter (Metric) Outer 26 mm
Overall Length (Metric) 255 mm
Packaging Quantity 4 Packs of 5
Product Type Digestion Tube with Socket