Learn More
Quickfit™ Borosilicate Glass Digestion Tube with Socket
Description
- Contains Quickfit precision-ground socket to accept air condensers
- Manufactured from borosilicate glass for highest chemical and thermal durability
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Diameter (Metric) Outer | 42 mm |
| Overall Length (Metric) | 300 mm |
| Packaging Quantity | 5/Pk. |
| Product Type | Digestion Tube with Socket |