Learn More
Fisherbrand™ Borosilicate Glass Conical Shape Narrow Neck Erlenmeyer Flasks
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
50 mL
500 mL
Unit Size:
Pack of 10
Description
Erlenmeyer, graduated.
Specifications
Specifications
| Certifications/Compliance | ISO 1773 |
| Diameter (Metric) Outer Neck | 34 mm |
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass |
| Shape | Conical |
| Mouth Style | Narrow |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido