Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Conical Separating Funnels with Solid Glass Plug, Scale
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Description
- With NS socket and PE stopper
- with solid glass plug
- with screwthread retaining device
- with scale (approx. volume /ml)
- material: borosilicate glass 3.3
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| Quantity | 2Pack |
| Stopper Material | Polyethylene |