Learn More
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Description
- Without scale; with solid glass plug
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Borosilicate Glass |
| Quantity | 2Pack |
| Stopper Material | Polyethylene |