Learn More
Borosilicate Glass Conical Form Beakers with Wide Mouth
Click to view available options
Capacity (Metric):
150 mL
250 mL
500 mL
Unit Size:
Each
Description
- Borosilicate Glass
- Conical
- Graduated
- Wide Mouth
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass |
| Includes | Spout |
| Quantity | 1 Ea. |
| Type | Beaker |