Learn More
BRAND™ BLAUBRAND™ Compact Automatic Burette with Plastic Bottle
Product Code. 10778785 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10778785 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10778785 Supplier BRAND™ Supplier No. 23821
  • ONLINE EXCLUSIVE PRICE
    1705.00 NOK
    1549.55 NOK / Pack of 1
    Save 155.45 NOK
    9% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Fast to dismantle and easy to clean. All individual components are replaceable.
  • Borosilicate glass (Boro 3.3) construction
  • Class AS, DE-M marking
  • Graduations in blue
  • Includes 1000 mL LDPE bottle with pedestal, PVC filling tube and detachable PTFE stopcock
  • DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
  • Supplied with a batch number and an accompanying batch certificate in the original packaging
  • On request, they are also available with an individual certificate, USP individual certificate or DAkkS calibration certificate
  • Support/clamp not included
  • TRUSTED_SUSTAINABILITY
    Specifications
    Color Clear
    Capacity (Metric) 50 mL
    Material Borosilicate Glass
    Graduations 0.1 mL
    Includes PE Bottle
    Stopcock Material PTFE
    Type Schellbach Buret
    Class Class AS
    Certifications/Compliance DIN EN ISO 385, USP, DIN EN ISO 385