Learn More
Kimble Chase™ Borosilicate Glass Class A Square Volumetric Flask
Click to view available options
Capacity (Metric):
500 mL
900 mL
Unit Size:
Each
Description
- Square cross-section with a width less than the widest diameter of a typical round flask
- All sizes cover 25-35% less area than conventional flasks
- Due to the added glass weight and the square section, flasks also have greater stability
- Graduation ring is blasted on the neck
- Calibrated to contain
- Calibrated to deliver
- With a marking spot
- Standard Taper ground glass stopper is supplied with the flask
- Designed from ASTM Specification E288, Class A requirements
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Calibration | Calibrated To Contain |
| Certifications/Compliance | ASTM Specification E288, Class A |
| Capacity (Metric) | 900 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 360 mm |
| No. per Case | 1 |
| Graduated | Yes |
| Class | A |
| Closure Type | Taper stopper |
| Closure Size | 22 |