Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Fritted Disc and PTFE Stopcock Plug
Description
- 40-60 micron porosity glass frit at the lower end to act as a support for column packing material
Specifications
Specifications
| For Use With (Application) | Ion exchange chromatography |
| Quantity | 1 Each |
| Diameter (Metric) Inner | 50 mm |
| Type | Chromatography Column |
| USP Type | Type I |
| Column Capacity | 589 mL |
| Length (Metric) | 300 mm |
| Material | Borosilicate Glass |
| Includes | With fritted disc and PTFE stopcock plug |