Learn More
Cytiva Borosilicate Glass Chromatographic Tubes
Product Code. 12477960 Shop All Cytiva Products
Click to view available options
Overall Length (Metric):
200 mm
40 cm
100 cm
For Use With:
10/40 Columns
16/100 Columns
For C 10/20 Columns
For C 16/20 Columns
Unit Size:
Each
Product Code. 12477960
Brand: Cytiva 19514001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Resistant to aqueous solutions and solvents used in liquid chromatography
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, > 10% HCl, > 5% acetic acid, or strong mineral acid.
TRUSTED_SUSTAINABILITY
Specifications
For Use With 16/100 Columns
Type Chromatographic Tubes
Overall Length (Metric) 100 cm
Material Borosilicate Glass
Maximum Pressure 14.5 psi
Min. Operating Temperature (Metric) 4°C
Max. Operating Temperature (Metric) 60°C