Learn More
Cytiva Borosilicate Glass Chromatographic Tubes
Shop All Cytiva Products
Click to view available options
Overall Length (Metric):
200 mm
40 cm
100 cm
For Use With:
10/40 Columns
16/100 Columns
For C 10/20 Columns
For C 16/20 Columns
Unit Size:
Each
Description
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, > 10% HCl, > 5% acetic acid, or strong mineral acid.
Specifications
Specifications
| For Use With | 10/40 Columns |
| Type | Chromatographic Tubes |
| Overall Length (Metric) | 40 cm |
| Material | Borosilicate Glass |
| Maximum Pressure | 14.5 psi |
| Min. Operating Temperature (Metric) | 4°C |
| Max. Operating Temperature (Metric) | 60°C |