Learn More
Cytiva Borosilicate Glass Chromatographic Tubes
Product Code. 11300122 Shop All Cytiva Products
Change view
Click to view available options
For Use With:
10/40 Columns
16/100 Columns
For C 10/20 Columns
For C 16/20 Columns
Unit Size:
Each
Overall Length (Metric):
200 mm
40 cm
100 cm
Product Code. For Use With unitSize Overall Length (Metric)
11300122 For C 16/20 Columns Each -
12477960 16/100 Columns Each 100 cm
11693184 10/40 Columns Each 40 cm
10231110 For C 10/20 Columns Each 200 mm
4 options
Product Code. 11300122
Brand: Cytiva 19513701
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Resistant to aqueous solutions and solvents used in liquid chromatography
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, > 10% HCl, > 5% acetic acid, or strong mineral acid.
TRUSTED_SUSTAINABILITY
Specifications
For Use With For C 16/20 Columns
Type Chromatography Tubes
Diameter (Metric) Inner 16 mm
Material Borosilicate Glass
Maximum Pressure 14.5 psi
Max. Operating Temperature (Metric) 60°C