Learn More
Cytiva Borosilicate Glass Chromatographic Tubes
Product Code. 10231110 Shop All Cytiva Products
Change view
Click to view available options
Overall Length (Metric):
200 mm
40 cm
100 cm
For Use With:
10/40 Columns
16/100 Columns
For C 10/20 Columns
For C 16/20 Columns
Unit Size:
Each
Product Code. Overall Length (Metric) For Use With unitSize
10231110 200 mm For C 10/20 Columns Each
12477960 100 cm 16/100 Columns Each
11693184 40 cm 10/40 Columns Each
11300122 - For C 16/20 Columns Each
4 options
Product Code. 10231110
Brand: Cytiva 19503601
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Resistant to aqueous solutions and solvents used in liquid chromatography
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, > 10% HCl, > 5% acetic acid, or strong mineral acid.
TRUSTED_SUSTAINABILITY
Specifications
For Use With For C 10/20 Columns
Type Chromatography Tubes
Diameter (Metric) Inner 10 mm
Overall Length (Metric) 200 mm
Material Borosilicate Glass
Maximum Pressure 14.5 psi
Max. Operating Temperature (Metric) 60°C