Description
- Withstands RCF up to 700.
- Borosilicate glass construction (Boro 3.3)
- Cylindrical with conical bottom
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| No. per Pack | 2 |
| Length (Metric) | 203 mm |
| Bottom Shape | Conical |
| Diameter (Metric) | 91 mm |
| Product Type | Centrifuge Tube |
| Max. RCF | 700 x g |