missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Supelco™

Product Code. 15813840
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15813840 1 kg 1kg
15893830 100 g 100g
15803840 500 g 500g
15823840 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15813840 Supplier Supelco Supplier No. 1001651000
  • ONLINE EXCLUSIVE PRICE
    960.00 NOK
    705.69 NOK / 1kg
    Save 254.31 NOK
    26% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS,ISO,Reag. Ph Eur

Boric acid (EMSURE™ grade product) is used for regulated analyses and highly demanding lab applications. It also offers the highest quality and unmatched scope of specifications to give complete control of test conditions and eliminate uncertainties. EMSURE™ grade products comply with all international regulations and can be used in various applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula H3BO3
Molecular Weight (g/mol) 61.83

Specifications

Melting Point 160°C (Dec.) (Lit.)
Density 1.489 g/cm3 at 23°C
Quantity 1 kg
MDL Number MFCD00011337
Chemical Name or Material Boric Acid
compliance-icons
Product Identifier
  • Boric acid for analysis EMSURE® ACS,ISO,Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to an approved waste disposal plant.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.