Learn More
JULABO™ Booster Heater 6 kW
The Booster Heater offers the option of increasing the heating capacity of some MAGIO laboratory circulators by 6 kW, thereby extending their operative range.
Brand: JULABO™ 9810007
Additional Details : Weight : 8.60000kg
Description
- Can be used in a temperature range from -50 to 300°C
- Intelligent control in combination with MAGIO ensures that it can be integrated into fully automated systems
Specifications
CE, UKCA | |
-50°C to +300°C | |
400 V, 50 Hz |
MAGIO Laboratory Circulator | |
Booster Heater |