Learn More
Berkshire International™ BCR™ Bond Paper
BIOPHARMA
Product Code. p-8011165 Shop All Berkshire Products
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Size:
A3
A4
A5
Product Type:
BCR Bond 1040
BCR Bond 680
BCR Bond 850
Unit Size:
Pack of 250
Product Code. Color Size Product Type unitSize
12847143 White A4 BCR Bond 680 Pack of 250
12857143 Blue A4 BCR Bond 680 Pack of 250
12897133 White A4 BCR Bond 850 Pack of 250
12807143 Blue A4 BCR Bond 850 Pack of 250
12837143 Green A4 BCR Bond 850 Pack of 250
12827143 Yellow A4 BCR Bond 850 Pack of 250
12817143 Pink A4 BCR Bond 850 Pack of 250
12877133 White A3 BCR Bond 850 Pack of 250
12887133 Blue A3 BCR Bond 850 Pack of 250
12867133 White A5 BCR Bond 1040 Pack of 250
12827133 White A4 BCR Bond 1040 Pack of 250
12837133 Blue A4 BCR Bond 1040 Pack of 250
12 options
Product Code. 12847143
Brand: Berkshire International™ B68A4WH25010
Only null left
Add to basket
Add to basket
Our BCR™ Bond is made with the purest quality cellulose from specially selected forests and is specially coated to minimize particle and fiber release.
  • Available in 5 colors: Green, Blue, Pink, White, and Yellow
cleanroom, paper, bond
Specifications
Material Cellulose
Color White
For Use With (Application) Writing, Copying, and Printing Within Controlled Environments
For Use With (Equipment) Photocopiers and Laser Printers
Page Format Sheets
Product Type BCR Bond 680
Size A4
Weight (Metric) 680 g