Description
- Available in 5 colors: Green, Blue, Pink, White, and Yellow
cleanroom, paper, bond
Specifications
Specifications
| Color | White |
| Description | Ultra-Clean |
| For Use With (Equipment) | Laser Printers and Photocopiers |
| Page Format | Sheets |
| Product Type | BCR Bond 1040 |
| Quantity | 250/Pk. |
| Size | A4 |