missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Triangular Magnetic Stirring Bars

PTFE-coated, triangular magnetic stirring bars.

Brand:  Bohlender™ C357-12

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 17137034

  • 253.34 NOK / Pack of 3
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

  • PTFE-encapsulated magnetic core (Alnico 5)
  • Universal chemical resistance
Specifications

Specifications

14 mm
0.55 in.
PTFE
Triangle
0.98 in.
Edge Length: 15 mm
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers