Learn More
Bohlender™ BOLA™ Tri-clamp Gaskets
Product Code. 17107344
Click to view available options
Diameter (Metric) Outside:
25 mm
34 mm
50.5 mm
64 mm
91 mm
For Use With (Equipment):
Tubing with 10 mm I.D.
Tubing with 16 mm I.D.
Tubing with 20 mm I.D.
Tubing with 26 mm I.D.
Tubing with 32 mm I.D.
Tubing with 38 mm I.D.
Tubing with 50 mm I.D.
Tubing with 66 mm I.D.
Size (Metric):
DN 10
DN 15
DN 20
DN 25
DN 32
DN 40
DN 50
DN 65
Unit Size:
Each
Product Code. 17107344
Brand: Bohlender™ D93024
Double-sided tri-clamp gaskets with elastic PTFE sleeve and silicone inlay.
  • Silicone ring with double-sided, elastic washer made of PTFE
  • Suitable for Tri-clamp connections as per DIN 32 676
  • Universal chemical resistance
  • Product is only exposed to PTFE
Specifications
For Use With (Equipment) Tubing with 16 mm I.D.
Size (Metric) DN 15
Diameter (English) Outside 1.3 in.
Diameter (Metric) Outside 34 mm
Material PTFE, Silicone