Learn More
Bohlender™ BOLA™ Reactor Lids for Glass Reactors DN 60
Product Code. 17186934
Click to view available options
Port Detail:
NPT 1/2 in. aside the shaft
NPT 1/4 in. to shaft, NPT 1/2 aside
Unit Size:
Each
Product Code. 17186934
Brand: Bohlender™ B15004
Reactor lids for glass reactors with Schott flat flange. With screw in threads for connection of transition fittings and a center neck NPT 1/2 in.
  • Suitable for glass reactors DN 60 with SCHOTT flat flange
  • With screw-in threads for connection of transition fittings and a center neck NPT 1/2 in.
  • Easy assembly
  • Flexibly expandable
  • Completely made of PTFE
  • Universal chemical resistance
  • With connectors for the use of existing equipment with ground joint or GL thread
Thread connections in detail:
  • Centric thread connection (purple) for insertion of a stirrer bearing
  • Vertical thread connection (blue) with parallel alignment to the stirrer shaft
  • Inclined thread connection (yellow) with direction straight to the stirrer shaft
  • Inclined thread connection (green) with direction aside the stirrer shaft
Specifications
Description Center Neck 1/2 in., Lateral Necks 3 x 1/2 in.
For Use With (Equipment) Glass Reactors DN 60 with SCHOTT Flat Flange
Product Type 4 x Female Thread Center Neck
Material PTFE
Port Detail NPT 1/2 in. aside the shaft