Learn More
Bohlender™ BOLA™ PTFE Tubing Connectors
Description
- PTFE straight fitting
- Two connectors for elastic tubing (e.g. Viton, Tygon, silicone)
- Universal chemical resistance
- Product is only exposed to PTFE
Compliance
FDA
Specifications
Specifications
| Certifications/Compliance | FDA |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 3 mm |
| Material | PTFE |
| Diameter (Metric) Outer | 6.8 mm |
| Overall Length (Metric) | 53 mm |
| For Use With | Elastic tubing |
| Min. Operating Temperature (Metric) | 0°C |
| Packaging Quantity | 10/Pk. |
| Tubing Size | 6.8mm O.D., 3mm I.D. |
| Show More |