Learn More
Bohlender™ BOLA™ PTFE Tubing, 0.79 mm Thick
Product Code. 17147604
Change view
Click to view available options
Diameter (English) Inner:
0.2 in.
Unit Size:
3m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Unit Size
17147604 0.2 in. 3m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17147604 Supplier Bohlender™ Supplier No. S181043
  • ONLINE EXCLUSIVE PRICE
    722.40 NOK
    662.73 NOK / 3m
    Save 59.67 NOK
    8% Off
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
Tubing made of virgin PTFE.
  • Translucent to milky-white appearance
  • Almost universal chemical resistance
  • Free from extractable agents
  • Physiologically safe
  • Non-adhesive surface
  • Very good sliding and dielectric characteristics
  • Flame retardant according to UL94V0
  • Oxygen value more than 95
  • Resistant to irradiation and weather
  • Can be sterilized in ETO
Specifications
Autoclavable Autoclavable
Diameter (English) Inner 0.2 in.
Diameter (Metric) Inner 4.78 mm
Material PTFE
Diameter (English) Outer 0.25 in.
Diameter (Metric) Outer 6.35 mm
Thickness (English) 0.03 in.
Thickness (Metric) 0.79 mm
Color Transparent
Flexibility 51 mm bend radius
Maximum Pressure 46 bar