Learn More
Bohlender™ Bola PTFE/PPS Ground Joint GL Adaptors
Product Code. 10422943
Change view
Click to view available options
Standard Taper Joint No.:
29/32
Unit Size:
Each
Product Code. Standard Taper Joint No. unitSize
10422943 29/32 Each
1 options
Product Code. 10422943
Brand: Bohlender™ D73440
Only null left
Add to basket
Add to basket
Assemble components with ground joints (condensers, stirrer bearings etc.) on glass bottles or GL-threaded necks with these adaptors. Bohlender™ BOLA™ Ground Joint GL Adaptors can be removed and fixed on another bottle without the risk of disarranging the tubing.
  • Black screw cap made of PPS with GL 45 thread
  • Movable insert with ground joint made of PTFE
  • Transition from a ground joint to a glass thread
  • The body can be turned independently from the screw cap
  • Very good chemical resistance
  • For working temperatures up to +250°C
Spécification
For Use With (Application) Assembly of components with ground joint (condensers, stirrer bearings etc.) on glass bottles or GL-threaded necks.
Material PPS, PTFE
Product Type Ground Joint GL Adaptors
Quantity 1 Each
Standard Taper Joint No. 29/32
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.