Learn More
Bohlender™ BOLA™ Measuring Cylinders
Product Code. 17196924
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
Graduations:
1 mL
2 mL
2.5 mL
5 mL
Diameter (Metric) Inside:
24 mm
30 mm
40 mm
53 mm
Height (Metric):
172 mm
202 mm
268 mm
308 mm
Unit Size:
Each
Product Code. Capacity (Metric) Graduations Diameter (Metric) Inside Height (Metric) unitSize
17196924 100 mL 2 mL 30 mm 202 mm Each
17186924 50 mL 1 mL 24 mm 172 mm Each
17106934 250 mL 2.5 mL 40 mm 268 mm Each
17116934 500 mL 5 mL 53 mm 308 mm Each
4 options
Product Code. 17196924
Brand: Bohlender™ A16416
Only null left
Add to basket
Add to basket
Translucent measuring cylinders with hexagonal base to prevent rolling.
  • Translucent and non-porous
  • Design based on ISO 4788
  • With graduation (uncalibrated), spout and reinforcing ring
  • Universal chemical resistance
  • Product is only exposed to PTFE
  • Hexagonal base prevents the cylinder from rolling
  • Permanent, recessed graduation
Specifications
Capacity (Metric) 100 mL
Material PTFE
Graduations 2 mL
Capacity (English) 3.9 in.
Shape Cylinder
Diameter (Metric) Inside 30 mm
Height (English) 8 in.
Height (Metric) 202 mm