Learn More
Bohlender™ BOLA™ Measuring Cylinders
Product Code. 17106934
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
Graduations:
1 mL
2 mL
2.5 mL
5 mL
Diameter (Metric) Inside:
24 mm
30 mm
40 mm
53 mm
Height (Metric):
172 mm
202 mm
268 mm
308 mm
Unit Size:
Each
Product Code. 17106934
Brand: Bohlender™ A16420
Only null left
Add to basket
Add to basket
Translucent measuring cylinders with hexagonal base to prevent rolling.
  • Translucent and non-porous
  • Design based on ISO 4788
  • With graduation (uncalibrated), spout and reinforcing ring
  • Universal chemical resistance
  • Product is only exposed to PTFE
  • Hexagonal base prevents the cylinder from rolling
  • Permanent, recessed graduation
Specifications
Capacity (Metric) 250 mL
Material PTFE
Graduations 2.5 mL
Capacity (English) 8.4 oz.
Shape Cylinder
Diameter (Metric) Inside 40 mm
Height (English) 11 in.
Height (Metric) 268 mm