Learn More
Bohlender™ BOLA™ Lids
Product Code. p-7237720
Change view
Click to view available options
For Use With (Equipment):
BOLA™ Beaker A136-03
BOLA™ Beaker A136-04
BOLA™ Beaker A136-05
Diameter (Metric) Outer:
27 mm
35 mm
40 mm
Diameter (English) Outer:
1.1 in.
1.4 in.
1.6 in.
Unit Size:
Pack of 2
Product Code. For Use With (Equipment) Diameter (Metric) Outer Diameter (English) Outer unitSize
17157384 BOLA™ Beaker A136-03 27 mm 1.1 in. Pack of 2
17167384 BOLA™ Beaker A136-04 35 mm 1.4 in. Pack of 2
17177384 BOLA™ Beaker A136-05 40 mm 1.6 in. Pack of 2
3 options
Product Code. 17157384
Brand: Bohlender™ H92703
Only null left
Add to basket
Add to basket
Tight-closing lids made of PTFE.
  • Lid with centering shoulder
  • Suitable for BOLA™™ Beakers
Specifications
Diameter (English) Outer 1.1 in.
Diameter (Metric) Outer 27 mm
Material PTFE
For Use With (Equipment) BOLA™ Beaker A136-03