Learn More
Bohlender™ BOLA™ Eco Stopcock Plugs with Bore, Taper 1:10, PP
Product Code. 30274462
Change view
Click to view available options
Plug Bore:
1.5 mm
2 mm
2.5 mm
4 mm
Closure Size:
NS 12.5 x 27 mm
NS 14.5 x 29 mm
NS 18.8 x 37 mm
For Use With (Equipment):
Thread M 10
Thread M 8
Unit Size:
Pack of 20
Pack of 3
Product Code. Plug Bore Closure Size For Use With (Equipment) unitSize
30274462 1.5 mm NS 12.5 x 27 mm Thread M 8 Pack of 20
17157484 1.5 mm NS 12.5 x 27 mm Thread M 8 Pack of 3
17167484 2 mm NS 12.5 x 27 mm Thread M 8 Pack of 3
30274469 2 mm NS 12.5 x 27 mm Thread M 8 Pack of 20
17187484 2.5 mm NS 14.5 x 29 mm Thread M 8 Pack of 3
30274460 2.5 mm NS 14.5 x 29 mm Thread M 8 Pack of 20
17107494 4 mm NS 18.8 x 37 mm Thread M 10 Pack of 3
30274493 4 mm NS 18.8 x 37 mm Thread M 10 Pack of 20
8 options
Product Code. 30274462
Brand: Bohlender™ J122003
Only null left
Add to basket
Add to basket
Eco stopcock plugs with nut, grip and washer made of polypropylene.
  • With bore
  • Smooth surface
  • Grip and nut made of polypropylene
  • Taper 1:10
  • Washer included
Specifications
Plug Bore 1.5 mm
Description Taper 1:10
Material Polypropylene
Closure Size NS 12.5 x 27 mm
For Use With (Equipment) Thread M 8
Color Red