Learn More
Bohlender™ BOLA™ Eco Stopcock Plugs with Bore, Taper 1:10, PP
Product Code. 17187484
Click to view available options
Plug Bore:
1.5 mm
2 mm
2.5 mm
4 mm
Closure Size:
NS 12.5 x 27 mm
NS 14.5 x 29 mm
NS 18.8 x 37 mm
For Use With (Equipment):
Thread M 10
Thread M 8
Unit Size:
Pack of 20
Pack of 3
Product Code. 17187484
Brand: Bohlender™ J122010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Eco stopcock plugs with nut, grip and washer made of polypropylene.
  • With bore
  • Smooth surface
  • Grip and nut made of polypropylene
  • Taper 1:10
  • Washer included
Specifications
Plug Bore 2.5 mm
Description Taper 1:10
Material Polypropylene
Closure Size NS 14.5 x 29 mm
For Use With (Equipment) Thread M 8
Color Red