Learn More
Bohlender™ BOLA™ Eco Stopcock Plugs with Bore, Taper 1:10, PP
Click to view available options
Plug Bore:
1.5 mm
2 mm
2.5 mm
4 mm
Closure Size:
NS 12.5 x 27 mm
NS 14.5 x 29 mm
NS 18.8 x 37 mm
For Use With (Equipment):
Thread M 10
Thread M 8
Unit Size:
Pack of 20
Pack of 3
Description
- With bore
- Smooth surface
- Grip and nut made of polypropylene
- Taper 1:10
- Washer included
Specifications
Specifications
| Plug Bore | 2 mm |
| Description | Taper 1:10 |
| Material | Polypropylene |
| Closure Size | NS 12.5 x 27 mm |
| For Use With (Equipment) | Thread M 8 |
| Color | Red |