Learn More
Bohlender™ BOLA™ Eco Stopcock Plugs with Bore, Taper 1:10, PP
Product Code. 17157484
Change view
Click to view available options
Plug Bore:
1.5 mm
2 mm
2.5 mm
4 mm
Closure Size:
NS 12.5 x 27 mm
NS 14.5 x 29 mm
NS 18.8 x 37 mm
For Use With (Equipment):
Thread M 10
Thread M 8
Unit Size:
Pack of 20
Pack of 3
Product Code. Plug Bore Closure Size For Use With (Equipment) unitSize
17157484 1.5 mm NS 12.5 x 27 mm Thread M 8 Pack of 3
30274462 1.5 mm NS 12.5 x 27 mm Thread M 8 Pack of 20
17167484 2 mm NS 12.5 x 27 mm Thread M 8 Pack of 3
30274469 2 mm NS 12.5 x 27 mm Thread M 8 Pack of 20
17187484 2.5 mm NS 14.5 x 29 mm Thread M 8 Pack of 3
30274460 2.5 mm NS 14.5 x 29 mm Thread M 8 Pack of 20
17107494 4 mm NS 18.8 x 37 mm Thread M 10 Pack of 3
30274493 4 mm NS 18.8 x 37 mm Thread M 10 Pack of 20
8 options
Product Code. 17157484
Brand: Bohlender™ J122003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Eco stopcock plugs with nut, grip and washer made of polypropylene.
  • With bore
  • Smooth surface
  • Grip and nut made of polypropylene
  • Taper 1:10
  • Washer included
Specifications
Plug Bore 1.5 mm
Description Taper 1:10
Material Polypropylene
Closure Size NS 12.5 x 27 mm
For Use With (Equipment) Thread M 8
Color Red