Learn More
Bohlender™ BOLA™ Balls
Description
- Made of solid PTFE, with smooth surface
- Different packing units
- For extension of surfaces such as distillation apparatus or as splash guard
Specifications
Specifications
| Material | PTFE |
| Diameter (Metric) | 3 mm |
| Description | Smooth Surface |
| Quantity | 100 Pk. |