Learn More
Bohlender™ BOLA™ Balls
Click to view available options
Diameter (Metric):
3 mm
9 mm
15 mm
25 mm
30 mm
Quantity:
100 Pk.
25 Pk.
50 Pk.
Unit Size:
Each
Pack of 25
Description
- Made of solid PTFE, with smooth surface
- Different packing units
- For extension of surfaces such as distillation apparatus or as splash guard
Specifications
Specifications
| Material | PTFE |
| Diameter (Metric) | 25 mm |
| Description | Smooth Surface |
| Quantity | 25 Pk. |