Learn More
Thermo Scientific Chemicals Boiling stones, for preparative work
Product Code. p-3586305 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. Quantity unitSize
10413375 250 g 250g
10340534 50 g 50g
2 options
Product Code. 10413375
Brand: Thermo Scientific Chemicals 386212500
Request Bulk or Custom Format
For preparative work
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Type Boiling Stone
Quantity 250 g