Learn More
Thermo Scientific Chemicals Boiling Stones, For Analytical Work
Product Code. p-3586302 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
Product Code. Quantity unitSize
10269473 50 g 50g
10219193 200 g 200g
2 options
Product Code. 10269473
Brand: Thermo Scientific Chemicals 386200500
Request Bulk or Custom Format
For analytical work
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Type Boiling Stone
Quantity 50 g