Learn More
Hanna Instruments BOILER WATER TEST KIT
Brand: Hanna Instruments HI3827
BOILER WATER TEST KIT
Safety and Handling
- Acid titrant solution / Diphenylcarbazone Indicator/ EDTA Reagent/ Mercuric Nitrate Solution / Nitric Acid Solution/ Phenolphtalein Indicator/ Phosphate Reagent/ Sulfamic Acid / Sulfuric Acid Solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H300-Fatal if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H332-Harmful if inhaled.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH071-Corrosive to the respiratory tract.
- EUH031-Contact with acids liberates toxic gas.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/first aider.
- P312-Call a POISON CENTER/doctor/first aider/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362-Take off contaminated clothing.
- P370+P378-In case of fire: Use to extinguish.
- P390-Absorb spillage to prevent material damage.
- P391-Collect spillage.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P404-Store in a closed container.
Supplemental information
- MIXTURE LIST-Contain: ethanolpotassium iodidee, sodium carbonate/hydrochloric acidmercury(II) nitrate,
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.