Learn More
Boc-N-methyl-L-alanine, TRC
Specifications
Specifications
| Chemical Name or Material | Boc-N-methyl-L-alanine |
| CAS | 16948-16-6 |
| Molecular Formula | C9H17NO4 |
| Quantity | 1 g |
| Synonym | N-[(1,1-Dimethylethoxy)carbonyl]-N-methyl-L-alanine, (S)-2-[(tert-Butoxycarbonyl)(methyl)amino]propanoic Acid, N-Methyl-N-(tert-butyloxycarbonyl)alanine, N-tert-Butoxycarbonyl-N-methylalanine |
| InChI Formula | InChI=1S/C9H17NO4/c1-6(7(11)12)10(5)8(13)14-9(2,3)4/h6H,1-5H3,(H,11,12)/t6-/m0/s1 |
| SMILES | C[C@H](N(C)C(=O)OC(C)(C)C)C(=O)O |
| IUPAC Name | (2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid |
| Molecular Weight (g/mol) | 203.24 |
| Formula Weight | 203.1158 |
Product Title
Haben Sie Verbesserungsvorschläge?