missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BMX, His Tag Recombinant Protein
Bone marrow tyrosine kinase (BMX) is a non-receptor tyrosine kinase that belongs to the SRC-related TEC subfamily of tyrosine kinases.
16000.00 NOK - 69300.00 NOK
Specifications
Accession Number | NP_001712 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 83 kDa |
Gene Alias | BMX, ETK, PSCTK3, PSCTK2 |
KinaseFamily | TEC Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15428433
|
Invitrogen™
PR5116C |
100μg |
16000.00 NOK
100µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10216192
|
Invitrogen™
PV4074 |
1 mg |
69300.00 NOK
1mg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
BMX is an important regulator of various cellular processes including apoptosis, cell survival, cellular differentiation, cell migration, and transformation.
Specifications
NP_001712 | |
83 kDa | |
TEC Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.05% Triton™ X-100, 4 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
BMX, ETK, PSCTK3, PSCTK2 | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Full Length, Recombinant, Cytoplasmic |