Learn More
BMS-935177, MedChemExpress
Product Code. 16490056
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Product Code. 16490056
Brand: Medchem Express HY101793
MedChemExpress BMS-935177 is a potent and selective reversible inhibitor of Bruton’s tyrosine kinase (Btk) with an IC50 of 3 nM.
BMS-935177 is a potent and selective reversible inhibitor of Bruton’s tyrosine kinase (Btk) with an IC50 of 3 nM.
Specifications
Chemical Name or Material BMS-935177
Physical Form Solid
CAS 1231889-53-4
Color Yellow
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C31H26N4O3
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 130 mg/mL (258.68 mM)
SMILES O=C(C1=CC=C(C2=CC=CC(N3C=NC4=C(C=CC=C4)C3=O)=C2C)C5=C1NC6=C5C=CC(C(C)(O)C)=C6)N
Molecular Weight (g/mol) 502.56
Formula Weight 502.56
Percent Purity 99.33%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.