Learn More
BMS-582949 hydrochloride, MedChemExpress
Product Code. p-200100918
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16496028 10 mg 10mg
16483416 25 mg 25mg
16468637 5 mg 5mg
16436446 100 mg 100mg
16402177 50 mg 50mg
5 options
Product Code. 16496028
Brand: Medchem Express HY14305A
MedChemExpress BMS-582949 hydrochloride is an orally active and highly selective p38α MAPK inhibitor, with an IC50 of 13 nM. BMS-582949 hydrochloride displays a significantly improved pharmacokinetic profile and is effective in inflammatory disease.
BMS-582949 hydrochloride is an orally active and highly selective p38α MAPK inhibitor, with an IC50 of 13 nM. BMS-582949 hydrochloride displays a significantly improved pharmacokinetic profile and is effective in inflammatory disease.
Specifications
Chemical Name or Material BMS-582949 hydrochloride
CAS 912806-16-7
Color White
Purity Grade Notes Research
Molecular Formula C22H27ClN6O2
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (56.44 mM; Need ultrasonic)
SMILES O=C(NCCC)C1=CN(N=CN=C2NC3=CC(C(NC4CC4)=O)=CC=C3C)C2=C1C.[H]Cl
Molecular Weight (g/mol) 442.94
Formula Weight 442.94
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation