Learn More
BMS-582949 hydrochloride, MedChemExpress
Product Code. 16496028
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16496028
Brand: Medchem Express HY14305A
MedChemExpress BMS-582949 hydrochloride is an orally active and highly selective p38α MAPK inhibitor, with an IC50 of 13 nM. BMS-582949 hydrochloride displays a significantly improved pharmacokinetic profile and is effective in inflammatory disease.
BMS-582949 hydrochloride is an orally active and highly selective p38α MAPK inhibitor, with an IC50 of 13 nM. BMS-582949 hydrochloride displays a significantly improved pharmacokinetic profile and is effective in inflammatory disease.
Specifications
Chemical Name or Material BMS-582949 hydrochloride
CAS 912806-16-7
Color White
Purity Grade Notes Research
Molecular Formula C22H27ClN6O2
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (56.44 mM; Need ultrasonic)
SMILES O=C(NCCC)C1=CN(N=CN=C2NC3=CC(C(NC4CC4)=O)=CC=C3C)C2=C1C.[H]Cl
Molecular Weight (g/mol) 442.94
Formula Weight 442.94
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.