missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMPR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19087-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody recognizes endogenous levels of total BMPR2 protein, including 115 kDa precursor, 75 kDa mature form and 150 kDa dimer.
This gene encodes a member of the bone morphogenetic protein (BMP) receptor family of transmembrane serine/threonine kinases. The ligands of this receptor are BMPs, which are members of the TGF-beta superfamily. BMPs are involved in endochondral bone formation and embryogenesis. These proteins transduce their signals through the formation of heteromeric complexes of two different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. Mutations in this gene have been associated with primary pulmonary hypertension, both familial and fenfluramine-associated, and with pulmonary venoocclusive disease.Specifications
BMPR2 | |
Polyclonal | |
Unconjugated | |
BMPR2 | |
BMP type 2 receptor, BMP type II receptor, BMPR 2, BMPR II, BMPR2, BMPR3, BMR2, BRK 3, PPH1, T ALK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12168, 140590, 659 | |
-20°C | |
Liquid |
Flow Cytometry, Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35607, Q13873 | |
Bmpr2 | |
BMPR2 Fusion Protein Ag5868 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |