Learn More
Brady™ BMP71 BMP61 M611 Vinyl General Identification Labels
Description
- Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Specifications
| Adhesive Type | Permanent Acrylic |
| Color | Green |
| Finish | Gloss |
| Material | Polyester (B-439) |
| Print Technology | Thermal Transfer |
| For Use With (Equipment) | BMP61, BMP71 (with Media Adapter), M611 |
| Dimensions (L x W) | 15.24 m x 25.4 mm |