Learn More
Brady™ BMP71 BMP61 M611 Vinyl General Identification Labels
Product Code. 11502545
Change view
Click to view available options
Dimensions (L x W):
15.24 m x 25.4 mm
15.24 m x 48.26 mm
Color:
Black
Blue
Green
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Color unitSize
11502545 15.24 m x 25.4 mm Blue Each
11512545 15.24 m x 48.26 mm Green Each
11522545 15.24 m x 48.26 mm Blue Each
11542545 15.24 m x 25.4 mm Black Each
11552545 15.24 m x 48.26 mm Black Each
11592535 15.24 m x 25.4 mm Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11502545 Supplier Brady™ Supplier No. 011179
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Labels for the BMP61 and M611 Thermal Transfer Portable Printers.
  • Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Adhesive Type Permanent Acrylic
Color Blue
Finish Gloss
Material Polyester (B-439)
Print Technology Thermal Transfer
For Use With (Equipment) BMP61, BMP71 (with Media Adapter), M611
Dimensions (L x W) 15.24 m x 25.4 mm