Learn More
Brady™ BMP61 M611 Self-Laminating Polyester Labels
A clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
Brand:  Brady™ 150853
Additional Details : Weight : 0.08300kg
 View more versions of this product
Product Code. 17150401
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Good smudge resistance, solvent resistance and high/low-temperature performance
  • Service temperature of -320 to 230° F (-196 to 110° C)
  • Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
Specifications
Specifications
Acrylic
Polyester
Printer Labels
BMP™61 Labeling System, BradyPrinter™ M611
1 x 1.75 in. (W x H)
1 in.
White
Thermal Transfer
Laboratory ID - Bottle, Slide, Tube & Vial
100/Pk.
0.0019 in.
1.75 in.